Learn More
Melford Nourseothricin Sulfate
Código de producto. p-200087194
Change view
Click to view available options
Cantidad:
100 mg
500 mg
1 g
Tamaño de la unidad:
1 g
100 miligramos
500 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30018588 1 g 1 g
30018586 100 mg 100 miligramos
30018587 500 mg 500 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30018588 Proveedor Melford N.º de proveedor N512001.0
Nourseothricin Sulfate is a broad-spectrum antibiotic.
Nourseothricin Sulfate is a broad-spectrum antibiotic. It is commonly used as a selection agent in molecular biology, genetics, and cell culture applications. Nourseothricin, also known as Streptothricin, is a natural product derived from Streptomyces species.
In molecular biology, Nourseothricin Sulfate is often employed as a selectable marker in gene expression studies and genetic engineering experiments. It works by inhibiting protein synthesis in bacterial and eukaryotic cells, leading to cell death.
Researchers use this antibiotic to introduce or maintain plasmids carrying resistance genes encoding Nourseothricin acetyltransferase (NAT) enzyme. The NAT enzyme inactivates Nourseothricin, allowing only the transformed cells to survive and propagate.
Nourseothricin Sulfate is also utilized in cell culture and microbiology. It can be used to selectively kill bacterial or fungal contaminants in cell cultures, thereby ensuring the purity of cell lines.
Additionally, it finds application in the isolation and maintenance of resistant strains in microbiological research.
Especificaciones
Nombre del producto químico o material Nourseothricin Sulfate
CAS 96736-11-7
Cantidad 1 g