missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOTCH2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marca:  Invitrogen™ PA527458

Código de producto. 13481407

  • 580.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Recommended positive controls: A431, HeLa, HepG2. Predicted reactivity: Mouse (95%), Rat (89%), Rhesus Monkey (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

NOTCH2
Polyclonal
Unconjugated
NOTCH2
AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2
Rabbit
Antigen affinity chromatography
RUO
18129, 4853
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
0.43 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
O35516, Q04721
NOTCH2
Recombinant protein encompassing a sequence within the Intracellular domain of human Notch2. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.