missing translation for 'onlineSavingsMsg'
Learn More

NOMO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Código de producto. 16807774 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Product Code. Cantidad unitSize
16807774 150 μL 150 microlitros
16817774 20 μL 20 microlitros
2 options
This item is not returnable. View return policy

Product Code. 16807774

Brand: Proteintech 177921AP150UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NOMO1 was originally thought to be related to the collagenase family. The gene encoding NOMO1 is one of three highly similar genes in a region of duplication located on the p arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development.

Specifications

Antígeno NOMO1
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.13 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica NOMO1
N.º de referencia del gen Q15155, Q6GQT9
Alias de gen NODAL modulator 1, Nomo, NOMO1, PM5, pM5 protein
Símbolos de los genes Nomo1
Especie del huésped Rabbit
Inmunógeno NOMO1 Fusion Protein Ag11895
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 211548, 23420, 361578
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.