Learn More
Invitrogen™ NOMO1 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Marca:  Invitrogen™ PA547534
Código de producto. 15588022
  • 572.00€ / 100 microgramos
Descripción
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a protein originally thought to be related to the collagenase gene family. This gene is one of three highly similar genes in a region of duplication located on the p arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development. Mutations in ABCC6, which is located nearby, rather than mutations in this gene are associated with pseudoxanthoma elasticum.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
NOMO1
Polyclonal
Unconjugated
NOMO1
D7Ertd156e; Nodal modulator 1; nodal modulator 3; Nomo; NOMO1; PM5; pM5 protein; pM5 protein 3; pM5 protein, telomeric copy; RGD1305240
Goat
Antigen affinity chromatography
RUO
23420
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q15155
NOMO1
Mouse myeloma cell line NS0-derived recombinant human NOMO Ser32-Asp1155.
100 μg
Primary
Human
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones