missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Antibody detects endogenous levels of total NLRC5.
NOD4 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD4 contains a CARD-like domain, a central NOD domain and a large LRR region. NOD4, an IFN-gamma-inducible nuclear protein, plays a role in homeostatic control of innate immunity and in antiviral defense mechanisms. As a key negative regulator of NF-kappa-B and type I interferon signaling, NOD4 may be a useful target for manipulating immune responses against infectious or inflammation-associated diseases, including cancer.
Especificaciones
Especificaciones
| Antígeno | NOD4 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide |
| génica | NLRC5 |
| N.º de referencia del gen | C3VPR6, Q86WI3 |
| Alias de gen | AI451557; AK220210; Caterpiller protein 16.1; CLR16.1; copine II; Cpne2; NLR family CARD domain containing 5; NLR family, CARD domain containing 5; Nlrc5; NOD27; NOD4; NOD-like receptor C5; Nucleotide-binding oligomerization domain protein 27; Nucleotide-binding oligomerization domain protein 4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 5; nucleotide-binding oligomerization domains 27; protein NLRC5 |
| Símbolos de los genes | NLRC5 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
