missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NME2 Monoclonal Antibody (4G7A8)

Descripción
MA1-185 detects NME2 from human samples. MA1-185 has been successfully tested in western blot and immunofluorescence applications. In western blot, MA1-185 detects NME2 (human) at the expected approximate molecular weight of 17kDa. The MA1-185 immunogen is full length NME2 expressed in E coli.
Nucleoside diphosphate kinase B is an enzyme that in humans is encoded by the NME2 gene. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by NME1) and 'B' (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein consisting of sequence sharing identity with each individual gene product.
Especificaciones
Especificaciones
| Antígeno | NME2 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 4G7A8 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.4 |
| génica | NME2 |
| N.º de referencia del gen | P19804, P22392, Q01768 |
| Alias de gen | 2.7.13.3; c-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; HEL-S-155an; histidine protein kinase NDKB; NDK B; NDKB; NDP kinase alpha; NDP kinase B; NDPKB; NDPK-B; nm23-2; NM23B; nm23-H2; nm23-M2; NME/NM23 nucleoside diphosphate kinase 2; Nme2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase B; P18; p18-12d; PUF |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?