missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMB Polyclonal Antibody, Invitrogen™
Description
NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.
Specifications
Specifications
| Antígeno | NMB |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | NMB |
| N.º de referencia del gen | P08949, Q9CR53 |
| Alias de gen | 3110023K12Rik; neuromedin B; neuromedin-B; Neuromedin-B-32; neuromedin-beta; NMB; RGD1562710 |
| Símbolos de los genes | NMB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?