Learn More
Novus Biologicals™ NM23-H2/NME2 Antibody (NME2/4160), Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals™ NBP3-26828-20ug
 Ver más versiones de este producto
Código de producto. 30043296
  • 232.00€ / 20 microgramos
Descripción
NM23-H2/NME2 Monoclonal antibody specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
NM23-H2/NME2
Monoclonal
0.2 mg/mL
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in
Recombinant full-length human NM23-H2/NME2 protein
20 μg
Primary
Human
Purified
Western Blot, Immunohistochemistry (Paraffin)
NME2/4160
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
4831
Store at 4°C.
IgG2 κ