missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nkx2.1 Recombinant Rabbit Monoclonal Antibody (JE61-73)
GREENER_CHOICE

Código de producto. 17806072
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
17806072 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17806072

Marca: Invitrogen™ MA544853

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

Nkx2.1 Recombinant Monoclonal Antibody for Western Blot, IHC (P), ICC/IF

This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Nkx2.1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon JE61-73
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
génica NKX2-1
N.º de referencia del gen P23441, P43699, P50220
Alias de gen AV026640; BCH; BHC; homeobox protein NK-2 homolog A; Homeobox protein Nkx-2.1; NK-2; NK2 homeobox 1; NK-2 homolog A; NKX2.1; Nkx-2.1; NKX2-1; NKX2A; NMTC1; RNA polymerase I transcription termination factor; T/EBP; TEBP; Thyroid nuclear factor 1; thyroid transcription factor 1; thyroid transcription factor 1 TTF-1 NK-2; thyroid transcription factor 1-like; thyroid-specific enhancer-binding protein; tinman; TITF1; TTF1; TTF-1
Símbolos de los genes NKX2-1
Especie del huésped Rabbit
Inmunógeno Recombinant protein within human TTF1 aa 272-371/371.
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21869, 25628, 7080
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.