Learn More
Nitrotyrosine Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems MAB3248-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Multi-Species samples. It is validated for Western Blot, Immunocytochemistry.
Especificaciones
| Nitrotyrosine | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Nitrotyrosine | |
| Nitrotyrosine-modified KLH | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
| Western Blot, Immunocytochemistry | |
| 306507 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects Nitrotyrosine adducts on proteins in Western blots. It does not cross-react with phosphotyrosine or 4-hydroxynonenal adducts. Unfixed cells, tissues, and proteins can be treated with Peroxynitrite (Catalog # AR006 ) for use as positive controls with this antibody. | |
| Multi-species | |
| Purified |
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido