Learn More
Lámina de Nitinol, 0,127 mm de grosor, memoria de forma, superficie plana recocida y decapada, Thermo Scientific Chemicals
Código de producto. p-7059870
Change view
Click to view available options
Cantidad:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Tamaño de la unidad:
cada uno
15423995 25 x 25 mm cada uno
15433995 50 x 50 mm cada uno
15443995 100 x 100 mm cada uno
3 options
Código de producto. 15423995
Marca: Thermo Scientific Alfa Aesar 045514.FF
Solicitar formato a granel o personalizado
Nitinol foil is used in cell phone antennas, children's toys, experimental materials, eye glasses components and medical equipment. It is also used in heat engines, resilient glass frames, mechanical watch springs, temperature control system, couplings, golf club insert, biomedical and medical uses, cell-phone technology: retractable antenna, microphone boom. It is highly biocompatible and has properties suitable for use in orthopaedic implants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Especificaciones
Nombre del producto químico o material Nitinol foil
Nombre de nota 0.127mm (0.005 in.) thick, shape memory, flat annealed, pickled surface
Cantidad 25 x 25 mm
Olor Odorless
Peso ≈0.51g/25x25mm
Número MDL MFCD02091734
Información de solubilidad Insoluble in water.