missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIRF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00€ - 468.00€
Especificaciones
| Antígeno | NIRF |
|---|---|
| Dilución | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
Descripción
NIRF Polyclonal antibody specifically detects NIRF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Especificaciones
| NIRF | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 115426 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp434B0920, DKFZp686G0837, E3 ubiquitin-protein ligase UHRF2, EC 6.3.2, EC 6.3.2.-, MGC33463, NIRFUbiquitin-like-containing PHD and RING finger domains protein 2, Np95/ICBP90-like RING finger protein, Np95-like RING finger protein, Nuclear protein 97, Nuclear zinc finger protein Np97, RNF107RING finger protein 107, Ubiquitin-like PHD and RING finger domain-containing protein 2, ubiquitin-like with PHD and ring finger domains 2, ubiquitin-like, containing PHD and RING finger domains, 2, URF2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 441-622 of human UHRF2 (NP_690856.1). IVPSNHYGPIPGIPVGSTWRFRVQVSEAGVHRPHVGGIHGRSNDGAYSLVLAGGFADEVDRGDEFTYTGSGGKNLAGNKRIGAPSADQTLTNMNRALALNCDAPLDDKIGAESRNWRAGKPVRVIRSFKGRKISKYAPEEGNRYDGIYKVVKYWPEISSSHGFLVWRYLLRRDDVEPAPWTS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto