Learn More
Invitrogen™ NIP7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA541863
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: VVVYSMADIP LGFGVAAKST QDCRKVDPMA IVVFHQADIG EYVRHEETLT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 93%; Yeast: 79%; Zebrafish: 79%.
NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
Especificaciones
| NIP7 | |
| Polyclonal | |
| Unconjugated | |
| Nip7 | |
| 1110017C15Rik; 60S ribosome subunit biogenesis protein NIP7 homolog; 60S ribosome subunit biogenesis protein NIP-like protein-like protein; 6330509M23Rik; AA408773; AA410017; CGI-37; FLJ10296; HSPC031; HSPC180; hypothetical protein LOC553616; KD93; kDa93; Nip7; NIP7 nucleolar pre-rRNA processing protein; NIP7, nucleolar pre-rRNA processing protein; Nip7p; nuclear import 7; nuclear import 7 homolog; nuclear import 7 homolog (S. cerevisiae); nucleolar pre-rRNA processing protein NIP7; OK/SW-cl.76; OK/SW-cl.78; pEachy; PUA protein; Saccharomyces cerevisiae Nip7p homolog; zgc:110384 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 51388 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y221 | |
| Nip7 | |
| Synthetic peptide directed towards the C-terminal of human NIP7 (aa 131-180). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |