missing translation for 'onlineSavingsMsg'
Learn More

Nintedanib, MedChemExpress

Código de producto. 16484047
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
1 g
10 miligramos
100 miligramos
200 miligramos
50 miligramos
500 miligramos
Product Code. Cantidad unitSize
16484047 50 mg 50 miligramos
16491326 200 mg 200 miligramos
16458138 10 mg 10 miligramos
16441936 500 mg 500 miligramos
16437896 100 mg 100 miligramos
16471949 1 g 1 g
6 options
This item is not returnable. View return policy

Product Code. 16484047

Brand: Medchem Express HY50904

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Nintedanib (BIBF 1120) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively.

Nintedanib (BIBF 1120) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively.

Specifications

Nombre del producto químico o material Nintedanib
CAS 656247-17-5
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C31H33N5O4
Cantidad 50 mg
Sinónimo BIBF 1120
Información de solubilidad DMSO : 11.5 mg/mL (21.31 mM; Need ultrasonic)
SMILES O=C1NC2=CC(C(OC)=O)=CC=C2/C1=C(NC3=CC=C(N(C(CN4CCN(C)CC4)=O)C)C=C3)\C5=CC=CC=C5
Peso molecular (g/mol) 539.62
Formula Weight (peso de la fórmula) 539.62
Porcentaje de pureza 98.52%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.