missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Niban-like protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA555049
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: FLQFYEDQYG VALFNSMRHE IEGTGLPQAQ LLWRKVPLDE RIVFSGNLFQ HQEDSKKWRN RFSLVPHNYG LVLYENKAAY ERQVPPRAVI NSAGYKILTS V Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
FAM129B (Family With Sequence Similarity 129 Member B, Niban Like Protein 1) is a Protein Coding gene. May play a role in apoptosis suppression. May promote melanoma cell invasion in vitro. Among its related pathways are MAP Kinase Signaling. An important paralog of this gene is FAM129A.
Especificaciones
| Niban-like protein | |
| Polyclonal | |
| Unconjugated | |
| Fam129b | |
| 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; family with sequence similarity 129 member B; family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; NIBAN2; Niban-like protein 1; OC58; Protein FAM129B; Protein Niban 2; RGD1307018 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64855 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96TA1 | |
| Fam129b | |
| Recombinant protein corresponding to Human Niban-like protein. Recombinant protein control fragment (Product #RP-92962). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido