Learn More
Invitrogen™ NHE3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marca:  Invitrogen™ PA542435
Código de producto. 15555462
  • 520.00€ / 100 microlitros
Descripción
Peptide sequence: AEDMVTHHTL QQYLYKPRQE YKHLYSRHEL TPTEDEKQDR EIFHRTMRKR Sequence homology: Cow: 77%; Dog: 92%; Human: 100%; Mouse: 92%; Pig: 77%; Rabbit: 77%; Rat: 92%; Sheep: 77%.
NHE3 (also known as SLC9A3 - Sodium/hydrogen exchanger 3) is a member of the sodium/hydrogen exchanger family that plays a crucial role in acid-base and volume homeostasis by mediating the majority of sodium and bicarbonate reabsorption in the proximal tubule of the kidney. Mutations in the gene can lead to Diarrhea 8.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
NHE3
Polyclonal
Unconjugated
SLC9A3
9030624O13Rik; AI930210; DIAR8; MGC126718; MGC126720; Na(+)/H(+) exchanger 3; Na/H exchanger 3; Na+/H+ exchanger homolog; Na+/H+ exchanger isoform 3; membrane ion exchange protein; Nhe3; NHE-3; NHE-3 antibody; plasma membrane ion transport protein; putative sodium hydrogen exchanger subtype 3; Slc9a3; SLC9A3 antibody; sodium proton exchanger; sodium/hydrogen exchanger; sodium/hydrogen exchanger 3; sodium/hydrogen exchanger isoform 3; Solute carrier family 9 (sodium/hydrogen exchanger 3), antiporter 3, Na+/H+ (amiloride insensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 3; solute carrier family 9 (sodium/hydrogen exchanger), member 3; solute carrier family 9 member 3; solute carrier family 9 member A3; solute carrier family 9, member 3; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3
Rabbit
Affinity chromatography
RUO
6550
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P48764
SLC9A3
Synthetic peptide directed towards the middle region of human SLC9A3.
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones