missing translation for 'onlineSavingsMsg'
Learn More

NGFR Monoclonal Antibody (OTI2A1), TrueMAB™, OriGene

Product Code. 17587594
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
Product Code. Cantidad unitSize
17587594 30 μL 30 microlitros
17567594 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 17587594

Brand: Origene Technologies TA812991S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NGFR Monoclonal antibody specifically detects NGFR in Human, Mouse, Rat samples. It is validated for Flow Cytometry

Specifications

Antígeno NGFR
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon OTI2A1
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica Ngfr
N.º de referencia del gen P07174, P08138, Q9Z0W1
Alias de gen CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human Ngfr (NP_002498) produced in HEK293T cell.
Método de purificación Protein A/G
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 18053, 24596, 4804
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.