missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFKBID Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA571595
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NFKBID Polyclonal Antibody for Western Blot, IHC (P)
May regulate the expression of IL-2, IL-6, and other cytokines through regulation on NF-kappa-B activity. Functions in the regulation of inflammatory responses. May also regulate TCR-induced negative selection of thymocytes.I-kappa-BNS, the nuclear I-kappa-B-like protein encoded by Nfkbid, is required for the development of marginal zone and peritoneal B-1 B cells and additionally required for extrafollicular antibody responses to T-independent and -dependent immunogens.I-kappa-BNS has been identified as a member of the I-kappa-B family of NF-kappa-B inhibitors, which undergoes induction upon TCR signaling. I-kappa-BNS mediates both positive and negative gene regulation, depending on individual cell type and/or cytokine.
Especificaciones
| NFKBID | |
| Polyclonal | |
| Unconjugated | |
| NFKBID | |
| ikappaBdelta; I-kappa-B-delta; IkappaBNS; IkB-delta; IKBNS; NF-kappa B inhibitor; NF-kappa-B inhibitor delta; NFKB inhibitor delta; Nfkbid; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, delta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, delta; TA-NFKBH; T-cell activation NFKB-like protein | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 243910, 84807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q2TB02, Q8NI38 | |
| NFKBID | |
| KLH conjugated synthetic peptide between 91-119 amino acids from the N-terminal region of human NFKBID. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido