missing translation for 'onlineSavingsMsg'
Learn More

NFKB1 Rabbit anti-Human, Polyclonal, Proteintech

Código de producto. 16842395 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Código de producto. Cantidad unitSize
16842395 20 μL 20 microlitros
16832395 150 μL 150 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16842395

Marca: Proteintech 235761AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody can bind p105 isoforms of NFKB1.

NF-kB is a nuclear transcription factor activated by various extra and intracellular stimuli such as cytokines, UV radiation, stress, in injury, and by bacterial and viral products. It is involved in regulation of various cellular events including cell growth, differentiation, proliferation, apoptosis and inflammation. NFKB1 (p50), a 50 kDa functional sub-unit of NF-kB, is a member of Rel protein family. It is synthesized as a p105 precursor protein and consists of an N-terminal conserved RHD-region containing nuclear localization signal, DNA-binding and dimerization domains. NFKB1 (p50) forms homodimers or heterodimerizes with p65, forming the functional NF-kB factor. NFKB1 (p50) directs the nuclear translocation of NF-kB and is instrumental in its DNA-binding. Pathological role of NF-kB has been suggested in AIDS, hematogenic cancer cell metastasis and rheumatoid arthritis.

Specifications

Antígeno NFKB1
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Clasificación Polyclonal
Concentración 0.54 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica NFKB1
N.º de referencia del gen P19838
Alias de gen DNA binding factor KBF1, EBP 1, KBF1, NF kappa B, NFKB p105, NFKB p50, NFKB1, NFKB1,p105, NFKB1,p105,p50, nfkb1a, p105, p50
Símbolos de los genes NFKB1
Especie del huésped Rabbit
Inmunógeno NFKB1 Fusion Protein Ag20297
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4790
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.