missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NF-H Monoclonal Antibody (RNF 402)
GREENER_CHOICE

Código de producto. 11540203
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
11540203 100 μL 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11540203

Marca: Invitrogen™ OMA106116

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

OMA1-06116 detects both the phosphorylated and non-phosphorylated forms of the 200 kDa neurofilament from rat, guinea pig, rabbit, dog, hamster, chicken, mouse, cow, monkey, sheep, xenopus and human tissues. OMA1-06116 has been successfully used in immunohistochemical (frozen & paraffin) and Western blot procedures. By Western blot, this antibody detects a single ~200 kDa protein representing both forms of the 200 kDa neurofilament from rat brain homogenate. OMA1-06116 antigen is cytoskeletal preparation of calf brain tissue.

Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno NF-H
Aplicaciones Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon RNF 402
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide
génica Nefh
N.º de referencia del gen P12036, P16884, P19246
Alias de gen 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg
Símbolos de los genes Nefh, nefh.L
Especie del huésped Mouse
Inmunógeno Neurofilament preparation of calf brain tissue.
Método de purificación Ammonium Sulfate Precipitation
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100101753, 100357177, 100716406, 100753127, 101107472, 101838568, 108715402, 24587, 380684, 417020, 442940, 4744, 528842
Especies diana Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Sheep, Hamster, Chicken, Guinea Pig, Frog
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgM
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.