Learn More
NeutraKine™ IL-3 Mouse anti-Human, Unconjugated, Clone: 3C11G7, Proteintech
Tienda Proteintech ProductosDescripción
The NeutraKine™ IL-3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-3. This antibody recognizes human antigen. The NeutraKine™ IL-3 antibody has been validated for the following applications: Neutralization, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | NeutraKine™ IL-3 |
| Aplicaciones | Neutralization |
| Clasificación | Monoclonal |
| Clon | 3C11G7 |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Cantidad | 25 μg |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 3562 |
| Especies diana | Human |
| Mostrar más |