Learn More
NeutraKine™ IFN-beta Mouse Monoclonal Antibody, Proteintech
Código de producto. p-200107384 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30237207 100 μg 100 microgramos
30236053 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30237207 Proveedor Proteintech N.º de proveedor 690131IG100UG
Mouse Monoclonal Antibody
The NeutraKine™ IFN beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™ IFN beta. This antibody recognizes human antigen. The NeutraKine™ IFN beta antibody has been validated for the following applications: ELISA, Neutralization analysis.
Especificaciones
Antígeno IFN beta
Aplicaciones ELISA, Neutralization
Clasificación Monoclonal
Clon 2C6E5
Conjugado Unconjugated
Símbolos de los genes IFB; IFF; IFNB; MGC96956
Especie del huésped Mouse
Inmunógeno RecombinantProtein
Método de purificación Protein G purification
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3456
Especies diana Human
Contenido y almacenamiento Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Tipo de producto Neu Antibody
Formulario Liquid
Isotype IgG1