Learn More
NeutraKine™ IFN Alpha 2A Mouse anti-Human, Unconjugated, Clone: 2H8G1, Proteintech
Tienda Proteintech ProductosDescripción
The NeutraKine™ IFN Alpha 2A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IFN Alpha 2A. This antibody recognizes human antigen. The NeutraKine™ IFN Alpha 2A antibody has been validated for the following applications: Neutralization, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | NeutraKine™ IFN Alpha 2A |
| Aplicaciones | Neutralization |
| Clasificación | Monoclonal |
| Clon | 2H8G1 |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Cantidad | 100 μg |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 3440 |
| Especies diana | Human |
| Mostrar más |