Learn More
Neuron-specific beta-III Tubulin Antibody, R&D Systems™
Código de producto. 18142971
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
18142971 100 μg 100 microgramos
1 options
Código de producto. 18142971
Marca: R&D Systems MAB1195
Mouse Monoclonal Antibody
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Multi-Species samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno beta-III Tubulin
Aplicaciones Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Clasificación Monoclonal
Clon TuJ-1
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 1-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.5-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Alias de gen Beta3-tubulin, beta-4, betaIII Tubulin, beta-Tubulin III, Class III beta-tubulin, TUBB3, TUBB4 CFEOM3A, Tubulin Beta 3, tubulin beta-3 chain, Tubulin beta-4, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Símbolos de los genes TUBB3
Especie del huésped Mouse
Inmunógeno Rat brain-derived microtubules
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10381
Especificidad de la prueba Detects mammalian and chicken neuron-specific beta-III tubulin but not other beta-tubulin isotypes in Western blots.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Multi-species
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2a