missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofascin Antibody, CoraFluor™ 1, Novus Biologicals™
Descripción
Neurofascin Polyclonal antibody specifically detects Neurofascin in Human samples. It is validated for ELISA
Especificaciones
Especificaciones
| Antígeno | Neurofascin |
| Aplicaciones | ELISA |
| Clasificación | Polyclonal |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | DKFZp686P2250, KIAA0756FLJ46866, neurofascin, neurofascin homolog, neurofascin homolog (chicken), NF, NRCAML |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with purified, recombinant Human Neurofascin (Accession#: O94856-12; Met1-Gln939) |
| Método de purificación | Antigen and protein A Affinity-purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?