missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroblastoma Antibody (FMG25), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-50528AF594
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Neuroblastoma Monoclonal specifically detects Neuroblastoma in Human samples. It is validated for Flow Cytometry.
Especificaciones
| Neuroblastoma | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Human cutaneous T cell lymphoma (HUT 78), SPF female Balb/c mice were injected i.p with 2 x 10*5 cells in complete Freund′s adjuvant. 30 days later a 2nd i.p injection of 2 x 10*5 cells without adjuvant was given. Fusion was 5 days | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| FMG25 | |
| Flow Cytometry | |
| Mouse | |
| Protein A purified | |
| Cancer | |
| Human | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Spot an opportunity for improvement?Share a Content Correction