Learn More
Neurexin 3/NRXN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-99886-100ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Neurexin 3/NRXN3 Polyclonal antibody specifically detects Neurexin 3/NRXN3 in Human samples. It is validated for ELISA
Especificaciones
| Neurexin 3/NRXN3 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| C14orf60, chromosome 14 open reading frame 60, KIAA0743MGC176711, neurexin 3, neurexin III, Neurexin III-alpha, Neurexin III-beta, neurexin-3-beta | |
| Produced in rabbits immunized with purified, recombinant Human Neurexin 3/NRXN3 (Accession#: NP_620426.2; Met1-Thr357) | |
| 100 μg | |
| Cell Biology, Neuroscience | |
| 9369 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |