missing translation for 'onlineSavingsMsg'
Learn More

NEU1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1F6D1, Proteintech

Product Code. 18068095 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Unit Size:
100 microlitros
20 microlitros
Product Code. Cantidad unitSize
18068095 20 μL 20 microlitros
18084755 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 18068095

Brand: Proteintech CL4886703220UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The NEU1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NEU1. This antibody recognizes human, rat, pig antigen. The NEU1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.

Specifications

Antígeno NEU1
Aplicaciones Immunofluorescence
Clon 1F6D1
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes Acetylneuraminyl hydrolase, FLJ93471, G9 sialidase, Lysosomal sialidase, N acetyl alpha neuraminidase 1, NANH, NEU, NEU1, NEURAMINIDASE 1, SIAL1, Sialidase 1
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 4758
Especies diana Human, Pig, Rat
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 155 kDa
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.