Learn More
Nestin Mouse, anti-Human, Unconjugated, Clone: 10C2, STEMCELL Technologies
Código de producto. 17188961
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17188961 100 μg 100 microgramos
17198961 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17188961 Proveedor Stemcell Technologies N.º de proveedor 60091
Mouse Monoclonal Antibody
  • The 10C2 antibody reacts with human nestin, a class VI intermediate filament (IF) protein expressed by embryonic neuroepithelial stem cells in the developing nervous system and by pancreatic islet and mesenchymal progenitor cells
  • Nestin expression is also observed in glioblastomas and many primary and metastatic melanomas and it is a useful angiogenic marker to evaluate neovascularity of endothelial cells in tumors
  • Nestin is required for the survival and renewal of neural progenitor cells and plays roles in cellular remodeling by forming heterodimers with vimentin or alpha-internexin, a process that appears to be tightly regulated by phosphorylation
  • Nestin localizes to the growth cones during axon elongation in differentiating neurons and may play a role in growth cone guidance
  • Nestin expression is down-regulated during differentiation, typically disappearing by embryonic stage E18
  • The 10C2 antibody reportedly cross-reacts weakly with monkey nestin but not rodent nestin
Especificaciones
Antígeno Nestin
Aplicaciones Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon 10C2
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 100 μg
Primario o secundario Primary
Especies diana Cynomolgus, Human
Isotype IgG1 κ
Tipo de célula Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Neurons