Learn More
Invitrogen™ NEFM Monoclonal Antibody (3H11)

Descripción
MA1-2011 detects the neurofilament, medium chain in human and rat samples. MA1-2011 has been successfully used in Western blot, immunofluorescent, and immunoprecipitation procedures. By Western blot this antibody detects a ~160 kDa protein representing the neurofilament, medium chain in HEK 293 cells, rat spinal cord, and human frontal cortex. In immunofluorecence procedures MA1-2011 recognizes the neurofilament, medium chain in human neurons. The MA1-2011 immunogen is a synthetic peptide corresponding to a C-terminal fragment of rat neurofilament.
Especificaciones
Especificaciones
| Antígeno | NEFM |
| Aplicaciones | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 3H11 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1mg/mL BSA and 0.05% sodium azide |
| génica | NEFM |
| N.º de referencia del gen | P07197, P12839 |
| Alias de gen | 160 kDa neurofilament protein; major neuronal intermediate filament subunit; medium neurofilament subunit; middle molecular weight neurofilament protein NF-M(2); Nef3; Nefm; nef-m; nefm.S; nefm-a; nefm-b; Neurofilament 3; neurofilament 3 (150kDa medium); neurofilament 3, medium; neurofilament M subunit; neurofilament medium; neurofilament medium polypeptide; neurofilament medium S homeolog; neurofilament medium tail domain; neurofilament protein M; Neurofilament protein, middle polypeptide; neurofilament triplet M protein; neurofilament, medium polypeptide; neurofilament, medium polypeptide 150kDa; neurofilament, medium polypeptide S homeolog; neurofilament-3 (150 kD medium); neurofilament-M; neurofilament-M subunit; neuronal intermediate filament; NF160; NF165; NFM; NF-M; NF-M c-terminus; NF-M protein; NMC; XELAEV_18020229mg |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.