Learn More
NEDD4 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Código de producto. 16851295 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16851295 20 μL 20 microlitros
16841295 150 μL 150 microlitros
2 options
Código de producto. 16851295
Marca: Proteintech 216981AP20UL
Rabbit Polyclonal Antibody
This antibody can recognize NEDD4-1 and NEDD4-2.
The E3 ubiquitin-protein ligase NEDD4 is a Ubl conjugation pathway protein. It contains one HECT (E6AP-type E3 ubiquitin-protein ligase) domain and four WW domains. It is involved in the pathway leading to the degradation of VEGFR-2/KDFR, independently of its ubiquitin-ligase activity. It acts as a potential proto-oncogene that negatively regulates PTEN via ubiquitination, a paradigm analogous to that of Mdm2 and p53. It is also reported to be involved in the budding of many viruses and is the regulator of epithelial Na+ channels, which lead to arterial hypertension. NEDD4 protein is localized to the sarcolemmal region of muscle fibers. Increase in NEDD4 protein expression leads to unloaded and denervated muscles as well as a decrease in Notch1 expression. Hence it acts as down regulator of Notch-1 gene expression.
Especificaciones
Antígeno NEDD4
Aplicaciones Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.22 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica NEDD4
N.º de referencia del gen P46934, P46935, Q62940
Alias de gen KIAA0093, NEDD 4, NEDD4, NEDD4 1, RPF1
Símbolos de los genes Nedd4
Especie del huésped Rabbit
Inmunógeno NEDD4 Fusion Protein Ag16291
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17999, 25489, 4734
Especies diana Mouse, Rat, Human, Non-human Primate
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG