Learn More
Invitrogen™ NDUFB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA529208
Descripción
Recommended positive controls: HepG2. Predicted reactivity: Rhesus Monkey (83%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Especificaciones
| NDUFB4 | |
| Polyclonal | |
| Unconjugated | |
| NDUFB4 | |
| 0610006N12Rik; 1300010H20Rik; B15; B15 subunit of the NADH: ubiquinone oxidoreductase complex; CI-B15; complex I B15 subunit; complex I-B15; MGC5105; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase subunit B4; NADH-ubiquinone oxidoreductase B15 subunit; Ndufb4; similar to B15 subunit of the NADH: ubiquinone oxidoreductase complex | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 288088, 4710 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95168 | |
| NDUFB4 | |
| Full length human NDUFB4 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.