Learn More
NCS1 Mouse anti-Human, Unconjugated, Clone: 2F10C8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67616-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The NCS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NCS1. This antibody recognizes human, mouse, rat, pig antigen. The NCS1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
| NCS1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2F10C8 | |
| Unconjugated | |
| 20 μL | |
| 23413 | |
| IgG2a |