Learn More
NCS1 Mouse anti-Human, Unconjugated, Clone: 2F10C8, Proteintech
Código de producto. 16810346 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16810346 150 μL 150 microlitros
16820346 20 μL 20 microlitros
2 options
Código de producto. 16810346
Marca: Proteintech 676161IG150UL
Mouse Monoclonal Antibody
The NCS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NCS1. This antibody recognizes human, mouse, rat, pig antigen. The NCS1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Antígeno NCS1
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence
Clasificación Monoclonal
Clon 2F10C8
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 150 μL
Primario o secundario Primary
ID de gen (Entrez) 23413
Especies diana Human, Mouse, Rat, Pig
Isotype IgG2a