Descripción
The NCK family of SH2/SH3 adaptor proteins consists of two members, NCK1 and NCK2, which couple tyrosine kinase signaling, including the EGF and PDGF receptor-pathways, to downstream signaling proteins. The NCK2 protein binds human PDGFR-b (Tyr-1009); overexpression of NCK2 inhibits PDGF-induced membrane ruffling and lamellipod formation.
Especificaciones
Especificaciones
| Antígeno | NCK2 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | NCK2 |
| N.º de referencia del gen | O43639, O55033 |
| Alias de gen | Cytoplasmic protein NCK2, GRB4, Nck 2, NCK adaptor protein 2, NCK2, NCKbeta |
| Símbolos de los genes | Nck2 |
| Mostrar más |