missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCAPH Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA528559
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.
Especificaciones
| NCAPH | |
| Polyclonal | |
| Unconjugated | |
| NCAPH | |
| A730011O11Rik; barren homolog 1; barren homolog protein 1; Brrn; BRRN1; Caph; CAP-H; Chromosome-associated protein H; condensin complex subunit 2; HCAP-H; KIAA0074; mCAP-H; mKIAA0074; Ncaph; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23397 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q15003 | |
| NCAPH | |
| Recombinant protein encompassing a sequence within the center region of human BRRN1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido