missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM1 Mouse anti-Human, Clone: UMAB83, lyophilized, UltraMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies UM500067CF
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD56 (NCAM, neural cell adhesion molecule) is a transmembrane glycoprotein of the immunoglobulin family that serves as an adhesive molecule and is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa. CD56 is found on T cells and NK cells, and is involved in cell migration, axonal growth, pathfinding and synaptic plasticity. Polysialic modification results in reduction of CD56-mediated cell adhesion. Through its extracellular region, CD56 mediates homophilic and heterophilic interactions by binding extracellular matrix components such as laminin and integrins. CD56 is expressed on most neuroectodermal derived cell lines, tissues and neoplasms such as retinoblastoma, medulloblastoma, astrocytomas and neuroblastoma. Further, CD56 is a widely used neuroendocrine marker with a high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms.Especificaciones
| NCAM1 | |
| Monoclonal | |
| Unconjugated | |
| NCAM1 | |
| CD56, MSK39, NCAM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4684 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Peptide Array | |
| UMAB83 | |
| PBS with 8% trehalose and no preservative | |
| P13591 | |
| NCAM1 | |
| Human recombit protein fragment corresponding to amino acids 20-718 of human NCAM1 produced in HEK293T cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido