Learn More
Invitrogen™ NARL Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5144474
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NARL Polyclonal Antibody for Western Blot, ELISA
This protein activates the expression of the nitrate reductase (narGHJI) and formate dehydrogenase-N (fdnGHI) operons and represses the transcription of the fumarate reductase (frdABCD) operon in response to a nitrate/nitrite induction signal transmitted by either the NarX or NarQ proteins.
Especificaciones
| NARL | |
| Polyclonal | |
| Unconjugated | |
| narL | |
| frdR | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 75203336 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.69 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P0AF28 | |
| narL | |
| Recombinant Escherichia coli (strain K12) NARL protein. | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |