missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthoquine phosphate, MedChemExpress
Descripción
Naphthoquine phosphate is a potent and orally active antimalarial agent. Naphthoquine phosphate has thorough killing function for various schizonts of plasmodia, including resistance of P. falciparum to Chloroquine.
Specifications
Specifications
| Nombre del producto químico o material | Naphthoquine phosphate |
| Forma física | Solid |
| CAS | 173531-58-3 |
| Color | Amarillo claro |
| Cantidad | 50 mg |
| Notas de grado de pureza | Research |
| Fórmula molecular | C24H34ClN3O9P2 |
| Para utilizar con (aplicación) | COVID-19-immunoregulation |
| Información de solubilidad | H2O : 3.33 mg/mL (5.50 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.65 mM; Need ultrasonic) |
| SMILES | ClC1=CC=C(C(NC2=C(CCCC3)C3=C(O)C(CNC(C)(C)C)=C2)=CC=N4)C4=C1.O=P(O)(O)O.O=P(O)(O)O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?