missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAMPT/PBEF Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Tienda Proteintech ProductosDescripción
Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.
Especificaciones
Especificaciones
| Antígeno | NAMPT/PBEF |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.33 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | NAMPT |
| N.º de referencia del gen | P43490, Q80Z29, Q99KQ4 |
| Alias de gen | 1110035O14Rik, NAmPRTase, NAMPT, PBEF, PBEF1, Pre B cell enhancing factor, VF, VISFATIN |
| Símbolos de los genes | Nampt, nampta |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?