Learn More
Thermo Scientific™ Tubos de polipropileno Nalgene™ 689, DI de 1/4
Realice una amplia variedad de aplicaciones con la excelente resistencia química a varios ácidos y álcalis de los tubos de polipropileno Thermo Scientific™ Nalgene™ 689.
en la oferta
Especificaciones
Certificaciones/conformidad Materiales de calidad alimentaria que cumplen la normativa 21 CFR 177.1520 sobre envasado de alimentos
Material Polipropileno
Longitud global (métrico) 30.48 m
Cantidad de embalaje 100
Ver más especificaciones
Descripción
Descripción
Perform a variety of applications with the excellent chemical resistance to several acids and alkalis of translucent and semirigid Thermo Scientific™ Nalgene™ 689 Polypropylene Tubing. This polypropylene tubing with durometer of 75 (Shore D) is unaffected by most solvents at ambient temperatures. Food-grade materials comply with 21 CFR 177.1520 for food packaging.
Features:
  • Should not be exposed to halogenated hydrocarbons for an extended period
  • Semirigid Durometer 75 (shore D)
  • Autoclavable
Ordering Information:
In packs of 100′ (30.5m)
Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.
Especificaciones
Especificaciones
Materiales de calidad alimentaria que cumplen la normativa 21 CFR 177.1520 sobre envasado de alimentos
Polipropileno
30.48 m
100
Documentos
Product Certifications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.