Learn More
Invitrogen™ MYST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA566837
Descripción
Immunogen sequence: EWVDKNRLALT KTVKDAVQKN SEKYLSELAE QPERKITRNQ KRKHDEINHV QKTYAEMDPT TAALEKEHE Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3), yeast YBF2 and SAS2, and TIP60 (HTATIP). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.
Especificaciones
| MYST1 | |
| Polyclonal | |
| Unconjugated | |
| KAT8 | |
| 2010203C02Rik; 5830450F21Rik; D7Ertd629e; h FLJ14040; Histone acetyltransferase KAT8; histone acetyltransferase Myst1; hMOF; K (lysine) acetyltransferase 8; K(lysine) acetyltransferase 8; Kat8; Lysine acetyltransferase 8; MOF; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; MYST histone acetyltransferase 1; MYST protein 1; MYST1; MYST-1; ortholog of Drosophila males absent on the first (MOF); PP7073; probable histone acetyltransferase MYST1; ZC2HC8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7Z6 | |
| KAT8 | |
| Recombinant Human MYST1. Recombinant protein control fragment (Product #RP-107492). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.