missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 11 Antibody (MYH11/923) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-48149
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Myosin heavy chain 11 Monoclonal specifically detects Myosin heavy chain 11 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| Myosin heavy chain 11 | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| MYH11 | |
| Recombinant full-length human Myosin heavy chain 11 protein (Uniprot: P35749) | |
| 7 mL | |
| Hypoxia, Stem Cell Markers | |
| 4629 | |
| Human, Rat | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| MYH11/923 | |
| Immunohistochemistry-Paraffin | |
| AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breast lesions and carcinoma in situ from invasive tumors. | |
| Store at 4C. | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido