missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYOCD Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA567810
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Predicted to react with Mouse and Rat samples.
Smooth muscle cells (SM) and cardiac muscle cells-specific transcriptional factor which uses the canonical single or multiple CArG boxes DNA sequence. Acts as a cofactor of serum response factor (SRF) with the potential to modulate SRF-target genes. Plays a crucial role in cardiogenesis and differentiation of the smooth muscle cell lineage (myogenesis).
Especificaciones
| MYOCD | |
| Polyclonal | |
| Unconjugated | |
| Myocd | |
| Basic SAP coiled-coil transcription activator 2; Bsac2; BSAC2A; Mycd; Myocardin; Myocd; SRF cofactor protein; SRF co-factor protein (cardiac and smooth muscle); Srfcp; transcription factor myocardin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 214384, 246297, 93649 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IZQ8, Q8R5I7, Q8VIM5 | |
| Myocd | |
| Synthesized peptide derived from human MYOCD. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction