Learn More
MYO9B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA124950
Descripción
PA1-24950 detects MYO9B from rat samples. PA1-24950 is expected to cross react with mouse and human due to sequence homology. PA1-24950 has been successfully used in Western blot procedures. The PA1-24950 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 1960-1980 of rat MYO9B. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
displays ATP-dependent binding to actin: binds members of the rho family of GTPases; may play a role in formation of the actin cytoskeleton.
Especificaciones
| MYO9B | |
| Polyclonal | |
| MYO9B | |
| MYO9B | |
| Synthetic peptide, conjugated to KLH, corresponding to residues 1960-1980 of rat MYO9B. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Q13459, Q9QY06, Q63358 | |
| Rabbit | |
| IgG fraction | |
| RUO | |
| 17925, 25486, 4650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.