Learn More
Invitrogen™ MYO6 Monoclonal Antibody (K70041_6D6), Invitrogen™
Description
Sequence of this protein is as follows: EEAERLRRIQ EEMEKERKRR EEDEKRRRKE EEERRMKLEM EAKRKQEEEE RKKREDDEKR IQAEVEAQLA RQKEEESQQQ AVLEQERRDR ELALRIAQSE AELISDEAQA DLALRRSLDS YPVSKNDGTR PKMTPEQMAK EMSEFLSRGP AVLATKAAAG TKKYDLSKWK YAELRDTINT SCDIELLAAC REEFHRRLKV YHAWKSKNKK RNTETEQRAP KSVTDYDFAP FLNNSPQQNP AAQIPARQRE IEMNRQQRFF RIPFIRPADQ YKDPQSKKKG WWYAHFDGPW IARQMELHPD KPPILLVAGK DDMEMCELNL EETGLTRKRG AEILPRQFEE
Specifications
Specifications
| Antígeno | MYO6 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Monoclonal |
| Clon | K70041_6D6 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| génica | MYO6 |
| N.º de referencia del gen | Q9UM54 |
| Alias de gen | BC029719; DFNA22; DFNB37; KIAA0389; MYO6; myosin; myosin VI; myosin-VI; RGD1560646; Snell's waltzer; sv; tailchaser; Tlc; unconventional myosin; unconventional myosin-6; Unconventional myosin-VI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.