Learn More
Invitrogen™ MYO1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA565795
Descripción
Immunogen sequence: FSLHLSEMSSV GSKGDFLLVS EHVIELLTKM YRAVLDATQR QLTVTVTEKF SVRFKENSVA VKVVQGPAGG DNSKLRY Highest antigen sequence identity to the following orthologs - mouse 82%, rat 78%.
The protein encoded by this gene belongs to the myosin superfamily. Myosins are molecular motors that, upon interaction with actin filaments, utilize energy from ATP hydrolysis to generate mechanical force. Each myosin has a conserved N-terminal motor domain that contains both ATP-binding and actin-binding sequences. Following the motor domain is a light-chain-binding 'neck' region containing 1-6 copies of a repeat element, the IQ motif, that serves as a binding site for calmodulin or other members of the EF-hand superfamily of calcium-binding proteins. At the C-terminus, each myosin class has a distinct tail domain that serves in dimerization, membrane binding, protein binding, and/or enzymatic activities and targets each myosin to its particular subcellular location. The kidney epithelial cell line, LLC-PK1-CL4 (CL4), forms a well ordered brush border (BB) on its apical surface. Experiments indicate that the brush border population of the encoded protein turns over rapidly, while its head and tail domains interact transiently with the core actin and plasma membrane, respectively. A rapidly exchanging pool of the protein encoded by this gene envelops an actin core bundle that, by comparison, is static in structure.
Especificaciones
| MYO1A | |
| Polyclonal | |
| Unconjugated | |
| MYO1A | |
| BBMI; BBM-I; brush border myosin 1; brush border myosin I; brush border myosin-I; DFNA48; MIHC; Myh1; MYHL; Myo1a; myosin heavy polypeptide 1 skeletal muscle adult; myosin I heavy chain; myosin IA; myosin, heavy polypeptide-like (100kD); myosin, heavy polypeptide-like (110kD); myosin-Ia; unconventional myosin-Ia | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4640 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBC5 | |
| MYO1A | |
| Recombinant Human MYO1A. Recombinant protein control fragment (Product #RP-106468). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.