Learn More
MYH10-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Código de producto. 18034515 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
18034515 20 μL 20 microlitros
18089685 100 μL 100 microlitros
2 options
Código de producto. 18034515
Marca: Proteintech CL4881967320UL
Rabbit Polyclonal Antibody
The MYH10-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH10-Specific. This antibody recognizes human, mouse, rat antigen. The MYH10-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Especificaciones
Antígeno MYH10-Specific
Aplicaciones Immunofluorescence
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes MYH10, Myosin 10, Myosin heavy chain 10
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 4628
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG