Learn More
MYH10-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Código de producto. 18089685 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18089685 100 μL 100 microlitros
18034515 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18089685 Proveedor Proteintech N.º de proveedor CL48819673100UL
Rabbit Polyclonal Antibody
The MYH10-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH10-Specific. This antibody recognizes human, mouse, rat antigen. The MYH10-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Especificaciones
Antígeno MYH10-Specific
Aplicaciones Immunofluorescence
Concentración 1000 μg/mL
Conjugado CoraLite Plus 488
Símbolos de los genes MYH10, Myosin 10, Myosin heavy chain 10
Especie del huésped Rabbit
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 4628
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG