missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MyD88 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA141072
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Suggested positive control: human spleen or RAW.
Myeloid differentiation Marker 88 (MyD88) is an adaptor molecule for the Toll/IL-1R and is involved in the inflammatory response induced by IL-1, IL-18 and LPS. MyD88 recruits IRAK to IL-1 receptor complex in response to IL-1. This pathway further leads to activation of NF-kB. MyD88 has a wide tissue distribution.
Especificaciones
| MyD88 | |
| Polyclonal | |
| Unconjugated | |
| MYD88 | |
| mutant myeloid differentiation primary response 88; Myd88; MYD88D; myeloid differentiation primary response 88; myeloid differentiation primary response gene (88); myeloid differentiation primary response gene 88; myeloid differentiation primary response protein MyD88 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17874, 301059, 420420, 444881, 4615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| A5HNF6, P22366, Q599T9, Q6Y1S1, Q99836 | |
| MYD88 | |
| A synthetic peptide corresponding to human MyD88 (between amino acids 200-270). | |
| 100 μg | |
| Primary | |
| Bovine, Chicken, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido